Learn More
enQuireBio™ Recombinant Mycoplasma 46 kDa surface antigen Protein
A DNA sequence encoding the Mycoplasma hyopneumoniae (strain 232) 46 kDa surface antigen, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7000-ye-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
WP_011206345.1 | |
Greater than 86.9% as determined by SDS-PAGE. | |
Research Use Only | |
p46 | |
Recombinant Protein | |
Yeast | |
CGQTESGSTS DSKPQAETLK HKVSNDSIRI ALTDPDNPRW ISAQKDIISY VDETEAATST ITKNQDAQNN WLTQQANLSP APKGFIIAPE NGSGVGTAVN TIADKGIPIV AYDRLITGSD KYDWYVSFDN EKVGELQGLS LAAGLLGKED GAFDSIDQMN EYLKSHMPQE TISFYTIAGS QDDNNSQYFY NGAMKVLKEL MKNSQNKIID LSPEGENAVY VPGWNYGTAG QRIQSFLTIN KDPAGGNKIK AVGSKPASIF KGFLAPNDGM AEQAITKLKL EGFDTQKIFV TGQDYNDKAK TFIKDGDQNM TIYKPDKVLG KVAVEVLRVL IAKKNKASRS EVENELKAKL PNISFKYDNQ TYKVQGKNIN TILVSPVIVT KANVDNPDA |
Mycoplasma 46 kDa surface antigen Protein | |
100 μg | |
Not determined. | |
Full Length | |
Mycoplasma | |
His | |
Tris-based buffer, 50% glycerol |