missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse NTF3 / Neurotrophin 3 Protein
Recombinant Protein
Marca: enQuireBio™ QP5425-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001157506.1 | |
NTF3 / Neurotrophin 3 Protein | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse NTF3 / Neurotrophin 3 induces Rat C6 cell proliferation. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
18205 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
NTF3 | |
Recombinant Protein | |
E. coli | |
MYAEHKSHRG EYSVCDSESL WVTDKSSAID IRGHQVTVLG EIKTGNSPVK QYFYETRCKE ARPVKNGCRG IDDKHWNSQC KTSQTYVRAL TSENNKLVGW RWIRIDTSCV CALSRKIGRT |