missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Lysyl oxidase homolog 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Lysyl oxidase homolog 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6316-ye-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_034859.2 | |
Lysyl oxidase homolog 1 Protein | |
Research Use Only | |
Loxl1 | |
Recombinant Protein | |
Yeast | |
RQAPSLPLPG RVGSDTVRGQ TRHPFGFGQV PDNWREVAVG DSTGMARART SVSQQRHGGS ASSSVSASAF ATTYRQPSPY PQQFPYPQAP FVNQYENYDP ASRTYEQGYV YYRGAGGGMG AGAAAVASAG VIYPFQPRAR YEDYGGGGGE EQPEYPAQGF YPAPERPYVP QPQPQPQPQP QPQPQPSDGL DRRYSHSLYN EGTPGFEQAY PDPSTDVSQA PAGAGGTYGG AGDPRLGWYP PYAANVPPEA YVPPRAVEPQ PPFRVLEPPY LPVRSSDAPS QGGERNGAQQ GRLSVGSVYR PNQNGRGLPD LVPDPNYVQA STYVQRAHLY SLRCAAEEKC LASTAYAPEA TDYDLRVLLR FPQRVKNQGT ADFLPNRPRH TWEWHSCHQH YHSMDEFSHY DLLDASTGKK VAEGHKASFC LEDSTCDFGN LKRYACTSHT QGLSPGCYDT YNADIDCQWI DITDVQPGNY ILKVHVNPKY IVLESDFTNN VVRCNIHYTG RYVSTTNCKI VQS | |
Greater than 90% as determined by SDS-PAGE. |
16949 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |