missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CCL3 / Mip1a Protein
Recombinant Protein
Marca: enQuireBio™ QP1450-500 ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_035467.1 | |
CCL3 / Mip1a Protein | |
Research Use Only | |
CCL3 | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
20302 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
APYGADTPTA CCFSYSRKIP RQFIVDYFET SSLCSQPGVI FLTKRNRQIC ADSKETWVQE YITDLELNA | |
Greater than 95% as determined by SDS-PAGE. |