missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse CCL17/TARC Protein

Código de producto. p-200055627 Tienda Bio Techne Productos
Click to view available options
Cantidad:
100 μg
1 mg
500 μg
5 μg
Unit Size:
1 miligramo
100 microgramos
5 microgramos
500 microgramos
This item is not returnable. View return policy

Product Code. 18451722

Brand: Novus Biologicals™ NBP2351831mg

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Recombinant Protein

TARC Protein is fully biologically active when compared to standard. The biologically active determined by a chemotaxis bioassay using human T-lymphocytes is in a concentration range of 1.0-10 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Para utilizar con (aplicación) PAGE, Bioactivity
Formulación Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
ID de gen (Entrez) 6361
Peso molecular M.W. (Theoretical): 7.9 kDa
Cantidad 1 mg
Requisitos de almacenamiento Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Concentración de endotoxinas Less than 1 EU/μg of CCL17/TARC as determined by LAL method.
Alias de gen ABCD-2, CC chemokine TARC, C-C motif chemokine 17, CCL17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARC/CCL17, TARCA-152E5.3, Thymus and activation-regulated chemokine
Símbolo de gen CCL17
Reconstitución Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C
Categoría de investigación Biologically Active Proteins, Chemokines and Cytokines
Grado de pureza o calidad >97%pure by SDS-PAGE and HPLC
Expresión CCL17/TARC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.