Descripción
A recombinant protein corresponding to CD80. Amino Acid Sequence: MGHTRRQGTSPSKCPYLNFFQLLVLAGLSHFCSGVIHVTKEVKEVATL SCGHNVSVEELAQTRIYWQKEKKMVLTMMSGDMNIWPEYKNRTIFDI TNNLSIVILALRPSDEGTYECVVLKYEKDAFKREHLAEVTLSVKADFPT PSISDFEIPTSNIRRIICSTSGGFPEPHLSWLENGEELNAINTTVSQDPETEL YAVSSKLDFNMTTNHSFMCLIKYGHLRVNQTFNWNTTKQEHFPDDKTHT CPPCPAPELLGGPSVFLFPPKPKDTLMISRTPEVTCVVVDVSHEDPEVKFNWYV DGVEVHNAKTKPREEQYNSTYRVVSVLTVLHQDWLNGKEYKCKV SNKALPAPIEKTISKAKGQPREPQVYTLPPSRDELTKNQVSLTCLVKGFYPSDI AVEWESNGQPENNYKTTPPVLDSDGSFFLYSKLTVDKSRWQQGNVFSCSVM HEALHNHYTQKSLSLSPGK The Recombinant Mouse B7-1/CD80 Protein has been validated for the following applications: SDS-Page, Cell depletion.
Especificaciones
Especificaciones
| ID de gen (Entrez) | 941 |
| Nombre | B7-1/CD80 Protein |
| Método de purificación | Protein A purified |
| Cantidad | 1 mg |
| Fuente | Baculovirus |
| Requisitos de almacenamiento | -80°C. Avoid freeze-thaw cycles. |