missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza-B Virus Florida 07/04 Protein
A cDNA sequence encoding the Influenza-B Virus Florida 07/04 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12438-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Influenza-B Virus Florida 07/04 Protein | |
Research Use Only | |
Recombinant Protein | |
Influenza B virus | |
The B/Florida/07/04 solution contains STE, 0.09% sodium azide (NaN3) and 0.005% thimerosal. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |