missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VHL Protein
A cDNA sequence encoding the VHL was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13934-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
VHL Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPRRAENWDE AEVGAEEAGV EEYGPEEDGG EESGAEESGPEESGPEELGA EEEMEAGRPR PVLRSVNSRE PSQVIFCNRS PRVVLPVWLN FDGEPQPYPT LPPGTGRRIH SYRGHLWLFR DAGTHDGLLV NQTELFVPSL NVDGQPIFAN ITLP | |
Greater than 95.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The Von Hippel-Lindau Protein contains 1x PBS pH 7.4, 2mM EDTA, and 1mM DTT. |