missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TXNDC17 / TRP14 / TXNL5 Protein
A DNA sequence encoding the Homo sapiens (Human) TXNDC17 / TRP14 / TXNL5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6858-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_116120.1 | |
TXNDC17 / TRP14 / TXNL5 Protein | |
Research Use Only | |
TXNDC17 | |
Recombinant Protein | |
E. coli | |
MARYEEVSVS GFEEFHRAVE QHNGKTIFAY FTGSKDAGGK SWCPDCVQAE PVVREGLKHI SEGCVFIYCQ VGEKPYWKDP NNDFRKNLKV TAVPTLLKYG TPQKLVESEC LQANLVEMLF SED | |
Greater than 90% as determined by SDS-PAGE. |
84817 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |