missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TSN Protein
A cDNA sequence encoding the TSN was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13828-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TSN Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSVSEIFVEL QGFLAAEQDI REEIRKVVQS LEQTAREILT LLQGVHQGAG FQDIPKRCLK AREHFGTVKT HLTSLKTKFP AEQYYRFHEH WRFVLQRLVF LAAFVVYLET ETLVTREAVT EILGIEPDRE KGFHLDVEDY LSGVLILASE LSRLSVNSVT AGDYSRPLHI STFINELDSG FRLLNLKNDS LRKRYDGLKY DVKKVEEVVY DLSIRGFNKE TAAACVEK | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The TSN protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 100mM NaCl, and 10% glycerol. |