Learn More
Novus Biologicals™ Recombinant Human TRBP Protein
Código de producto. 18131649 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0,1 mg
0,5 mg
Tamaño de la unidad:
0.10 miligramo
0.50 miligramo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18131649 0,5 mg 0.50 miligramo
18146627 0,1 mg 0.10 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18131649 Proveedor Novus Biologicals™ N.º de proveedor NBP2517140.5MG
Highly purified. Generating reliable and reproducible results.
An un-tagged denatured recombinant protein corresponding to the amino acids 1-345 of Human TRBP The Recombinant Human TRBP Protein is derived from E. coli. The Recombinant Human TRBP Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Especificaciones
Concentración 1mg/mL
Para utilizar con (aplicación) SDS-PAGE
Formulación 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol 0.4M UREA
ID de gen (Entrez) 6895
Peso molecular TMW: 36.9kDa
Cantidad 0,5 mg
Requisitos de almacenamiento Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Alias de gen LOQS, RISC-loading complex subunit TARBP2, TAR (HIV) RNA binding protein 2, TAR (HIV) RNA-binding protein 2, TAR (HIV) RNA-binding protein TRBP1, TAR (HIV-1) RNA binding protein 2, TAR RNA binding protein 2, TAR RNA-binding protein 2, trans-activation responsive RNA-binding protein, Trans-activation-responsive RNA-binding protein, TRBP, TRBP1, TRBP2
Nombre común TRBP
Conjugado Unconjugated
Recombinante Recombinant
Grado de pureza o calidad >85%, by SDS-PAGE
Mostrar más Mostrar menos
For research use only and is not approved for use in humans or in clinical diagnosis
Título del producto
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.