missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcobalamin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcobalamin-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6769-ye-100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001053.2 | |
Transcobalamin-1 Protein | |
Research Use Only | |
TCN1 | |
Recombinant Protein | |
Yeast | |
EICEVSEENY IRLKPLLNTM IQSNYNRGTS AVNVVLSLKL VGIQIQTLMQ KMIQQIKYNV KSRLSDVSSG ELALIILALG VCRNAEENLI YDYHLIDKLE NKFQAEIENM EAHNGTPLTN YYQLSLDVLA LCLFNGNYST AEVVNHFTPE NKNYYFGSQF SVDTGAMAVL ALTCVKKSLI NGQIKADEGS LKNISIYTKS LVEKILSEKK ENGLIGNTFS TGEAMQALFV SSDYYNENDW NCQQTLNTVL TEISQGAFSN PNAAAQVLPA LMGKTFLDIN KDSSCVSASG NFNISADEPI TVTPPDSQSY ISVNYSVRIN ETYFTNVTVL NGSVFLSVME KAQKMNDTIF GFTMEERSWG PYITCIQGLC ANNNDRTYWE LLSGGEPLSQ GAGSYVVRNG ENLEVRWSKY | |
Greater than 90% as determined by SDS-PAGE. |
6947 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |