missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Especificaciones
Especificaciones
| Para utilizar con (aplicación) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulación | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| ID de gen (Entrez) | 2475 |
| Peso molecular | MolecularWeight-theroretical: 36.63 kDa |
| Cantidad | 10 μg |
| Requisitos de almacenamiento | Store at -80°C. Avoid freeze-thaw cycles. |
| Alias de gen | EC 2.7.11.1, FK506 binding protein 12-rapamycin associated protein 1, FK506 binding protein 12-rapamycin associated protein 2, FK506-binding protein 12-rapamycin complex-associated protein 1, FKBP12-rapamycin complex-associated protein, FLJ44809, FRAP, FRAP1FKBP12-rapamycin complex-associated protein 1, FRAP2, Mammalian target of rapamycin, Mechanistic target of rapamycin, mechanistic target of rapamycin (serine/threonine kinase), mTOR, RAFT1, rapamycin and FKBP12 target 1, rapamycin associated protein FRAP2, Rapamycin target protein 1, RAPT1FKBP-rapamycin associated protein, serine/threonine-protein kinase mTOR |
| Símbolo de gen | MTOR |
| Categoría de investigación | Autophagy, Cancer, Cell Cycle and Replication, DNA Repair, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Signal Transduction, Translation Control |
| Grado de pureza o calidad | >80% by SDS-PAGE and Coomassie blue staining |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?