missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TMEM27 Protein
A cDNA sequence encoding the TMEM27 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13763-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TMEM27 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASELCQPGAENA FKVRLSIRTA LGDKAYAWDT NEEYLFKAMV AFSMRKVPNR EATEISHVLL CNVTQRVSFW FVVTDPSKNH TLPAVEVQSA IRMNKNRINN AFFLNDQTLE FLKIPSTLAP PMDPSVP | |
Greater than 95.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TMEM27 filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH 4.0. |