missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human SPDYE2 GST (N-Term) Protein

Product Code. 18585970 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
2 μg
Unit Size:
2 microgramos
Product Code. Cantidad unitSize
18585970 2 μg 2 microgramos
1 options
This item is not returnable. View return policy

Product Code. 18585970

Brand: Novus Biologicals™ H00441273P012ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

Para utilizar con (aplicación) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulación 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
ID de gen (Entrez) 441273
Peso molecular MolecularWeight-theroretical: 58.2 kDa
Cantidad 2 μg
Requisitos de almacenamiento Store at -80°C. Avoid freeze/thaw cycles.
Alias de gen MGC119295, putative Speedy protein E2, speedy homolog E2 (Xenopus laevis), speedy protein E2
Símbolo de gen SPDYE2
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Expresión SPDYE2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.