missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RELA / Transcription factor p65 / NFkB p65 Protein
A DNA sequence encoding the Homo sapiens (Human) RELA / Transcription factor p65 / NFkB p65, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP1321-ec-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001138610.1 | |
RELA / Transcription factor p65 / NFkB p65 Protein | |
Research Use Only | |
RELA | |
Recombinant Protein | |
E. coli | |
MDELFPLIFP AEPAQASGPY VEIIEQPKQR GMRFRYKCEG RSAGSIPGER STDTTKTHPT IKINGYTGPG TVRISLVTKD PPHRPHPHEL VGKDCRDGFY EAELCPDRCI HSFQNLGIQC VKKRDLEQAI SQRIQTNNNP FQVPIEEQRG DYDLNAVRLC FQVTVRDPSG RPLRLPPVLS HPIFDNRAPN TAELKICRVN RNSGSCLGGD | |
Greater than 90% as determined by SDS-PAGE. |
5970 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |