missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Regulator of microtubule dynamics protein 1
A DNA sequence encoding the Homo sapiens (Human) Regulator of microtubule dynamics protein 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7878-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001291731.1 | |
Regulator of microtubule dynamics protein 1 | |
Research Use Only | |
RMDN3 | |
Recombinant Protein | |
E. coli | |
MSRLGALGGA RAGLGLLLGT AAGLGFLCLL YSQRWKRTQR HGRSQSLPNS LDYTQTSDPG RHVMLLRAVP GGAGDASVLP SLPREGQEKV LDRLDFVLTS LVALRREVEE LRSSLRGLAG EIVGEVRCHM EENQRVARRR RFPFVRERSD STGSSSVYFT ASSGATFTDA ESEGGYTTAN AESDNERDSD KESEDGEDEV SCETVKMGRK DSLDLEEEAA SGASSALEAG GSSGLEDVLP LLQQADELHR GDEQGKREGF QLLLNNKLVY GSRQDFLWRL ARAYSDMCEL TEEVSEKKSY ALDGKEEAEA ALEKGDESAD CHLWYAVLCG QLAEHESIQR RIQSGFSFKE HVDKAIALQP ENPMAHFLLG RWCYQVSHLS WLEKKTATAL LESPLSATVE DALQSFLKAE ELQPGFSKAG RVYISKCYRE LGKNSEARWW MKLALELPDV TKEDLAIQKD LEELEVILRD | |
Greater than 90% as determined by SDS-PAGE. |
55177 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |