missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human PRAC GST (N-Term) Protein
Tienda Bio Techne Productos
missing translation for 'orderingAttributeHoverText'
Cantidad:
10 μg
25 μg
Förpackningsstorlek
10 microgramos
25 microgramos
Beskrivning
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifikationer
Specifikationer
| Para utilizar con (aplicación) | Western Blot, ELISA, SDS-PAGE, Protein Array, Immunoaffinity Purification |
| Formulación | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| ID de gen (Entrez) | 84366 |
| Peso molecular | MolecularWeight-theroretical: 32.4 kDa |
| Cantidad | 10 μg |
| Requisitos de almacenamiento | Store at -80°C. Avoid freeze/thaw cycles. |
| Alias de gen | C17orf92, chromosome 17 open reading frame 92, MGC32520, PRAC1, prostate cancer susceptibility candidate, Prostate, rectum and colon expressed gene protein, small nuclear protein PRAC |
| Símbolo de gen | PRAC1 |
| Categoría de investigación | Cancer, Cell Biology, Tumor Suppressors |
| Grado de pureza o calidad | >80% by SDS-PAGE and Coomassie blue staining |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering