Learn More
enQuireBio™ Recombinant Human PIGX Protein
A DNA sequence encoding the Homo sapiens (Human) PIGX, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7951-ec-1mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001159776.1 | |
PIGX Protein | |
Research Use Only | |
PIGX | |
Recombinant Protein | |
E. coli | |
MCSEIILRQE VLKDGFHRDL LIKVKFGESI EDLHTCRLLI KQDIPAGLYV DPYELASLRE RNITEAVMVS ENFDIEAPNY LSKESEVLIY ARRDSQCIDC FQAFLPVHCR YHRPHSEDGE ASIVVNNPDL LMFCDQEFPI LKCWAHSEVA APCALENEDI CQWNKMKYKS VYKNVILQVP VGLTVHTSL | |
Greater than 90% as determined by SDS-PAGE. |
54965 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |