Learn More
Novus Biologicals™ Recombinant Human NTNG1 GST (N-Term) Protein
Código de producto. 18351989 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
10 μg
25 μg
Tamaño de la unidad:
10 microgramos
25 microgramos
18351989 10 μg 10 microgramos
18335019 25 μg 25 microgramos
2 options
Código de producto. 18351989
Marca: Novus Biologicals™ H00022854P0110ug
Recombinant Protein
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY
Especificaciones
Para utilizar con (aplicación) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulación 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
ID de gen (Entrez) 22854
Peso molecular 70.84 kDa
Cantidad 10 μg
Requisitos de almacenamiento Store at -80°C. Avoid freeze-thaw cycles.
Alias de gen axon guidance molecule, KIAA0976YLSR571, laminet 1, laminet-1, Lmnt1, netrin G1, netrin G1f, netrin-G1
Símbolo de gen NTNG1
Categoría de investigación Neuroscience
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Expresión NTNG1