missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NTF4 Protein
A DNA sequence encoding the Human NTF4, was expressed in the hosts and tags indicated.
105.00€ - 3420.00€
Especificaciones
Número de acceso | NP_006170.1 |
---|---|
ID de gen (Entrez) | 4909 |
Nombre | NTF4 Protein |
Estado normativo | Research Use Only |
Concentración de endotoxinas | Less than 1.0 EU/ug as determined by LAL method. |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15923778
|
enQuireBio™
QP10412-ec-2ug |
2 μg | |||||||
15903778
|
enQuireBio™
QP10412-ec-10ug |
10 μg | |||||||
15993768
|
enQuireBio™
QP10412-ec-100ug |
100 μg | |||||||
15913778
|
enQuireBio™
QP10412-ec-250ug |
250 μg | |||||||
Especificaciones
NP_006170.1 | |
NTF4 Protein | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MGVSETAPAS RRGELAVCDA VSGWVTDRRT AVDLRGREVE VLGEVPAAGG SPLRQYFFET RCKADNAEEG GPGAGGGGCR GVDRRHWVSE CKAKQSYVRA LTADAQGRVG WRWIRIDTAC VCTLLSRTGR A | |
Greater than 97% as determined by SDS-PAGE and HPLC. |
4909 | |
Research Use Only | |
NTF4 | |
The ED50 as determined by the dose-dependent induction of choline acetyl transferase activity in rat basal forebrain primary septal cell cultures is less than 50 ng/ml, corresponding to a specific activity of > 2.0 M 104 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 5.5, lyophilized |