missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human NSMAF GST (N-Term) Protein

Código de producto. 18668700 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
10 μg
25 μg
Tamaño de la unidad:
10 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18668700 10 μg 10 microgramos
18618840 25 μg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18668700 Proveedor Novus Biologicals™ N.º de proveedor H00008439Q0110ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Especificaciones

Para utilizar con (aplicación) Western Blot, ELISA, SDS-PAGE, Protein Array, Immunoaffinity Purification
Formulación 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
ID de gen (Entrez) 8439
Peso molecular MolecularWeight-theroretical: 36.74 kDa
Cantidad 10 μg
Requisitos de almacenamiento Store at -80°C. Avoid freeze-thaw cycles.
Alias de gen Factor associated with neutral sphingomyelinase activation, FANFactor associated with N-SMase activation, neutral sphingomyelinase (N-SMase) activation associated factor, protein FAN
Símbolo de gen NSMAF
Categoría de investigación Cell Biology, Immunology, Innate Immunity, Signal Transduction
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Expresión NSMAF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.