Especificaciones
| Concentración | 0.25 mg/mL |
| Para utilizar con (aplicación) | SDS-PAGE |
| Formulación | PBS (pH 7.4), 10% glycerol |
| ID de gen (Entrez) | 3821 |
| Peso molecular | MolecularWeight-theroretical: 17.1 kDa |
| Cantidad | 10 μg |
| Requisitos de almacenamiento | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Concentración de endotoxinas | Less Than 1.0 EU / 1 μg of protein (determined by LAL method) |
| Alias de gen | CD159 antigen-like family member A, CD159A, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B |
| Símbolo de gen | KLRC1 |
| Mostrar más |