missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Neuregulin-1 beta 1/NRG1 beta 1 Protein

Produktkod. 18459491 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Cantidad:
100 μg
10 μg
1 mg
500 μg
Förpackningsstorlek:
1 miligramo
10 microgramos
100 microgramos
500 microgramos
Produktkod. Cantidad unitSize
18459491 500 μg 500 microgramos
18427261 100 μg 100 microgramos
18476711 10 μg 10 microgramos
18410152 1 mg 1 miligramo
4 options
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 18459491

Brand: Novus Biologicals™ NBP235097500ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

Neuregulin-1 beta 1/NRG1 beta 1 protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using serum free human MCF-7 cells is less than 0.5 ng/mL, corresponding to a specific activity of >2.0 x 106IU/mg.
TRUSTED_SUSTAINABILITY

Specifikationer

Número de acceso Q02297, Q02297, Q02297
Concentración LYOPH
Para utilizar con (aplicación) PAGE, Bioactivity
Formulación Lyophilized from a 0.2 um filtered solution in 1 x PBS, pH 7.4.
ID de gen (Entrez) 3084
Peso molecular 7.5 kDa
Cantidad 500 μg
Requisitos de almacenamiento Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Concentración de endotoxinas Less than 0.1 EU/ug of Neuregulin-1 beta 1/NRG1 beta 1 as determined by LAL method.
Alias de gen ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, MSTP131, NDF, neuregulin 1, NRG1-IT2, SMDF
Símbolo de gen NRG1
Reconstitución Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Grado de pureza o calidad >97% pure by SDS-PAGE and HPLC
Expresión Neuregulin-1 beta 1/NRG1 beta 1
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.