missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MICB Protein
A DNA sequence encoding the Human MICB, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10173-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001276089.1 | |
MICB Protein | |
Research Use Only | |
MICB | |
The specific activity is determined by binding MICB antibody in ELISA. | |
Human | |
Untagged | |
0.2 um filtered 20 mM Tris, 150 mM NaCl, pH 8.0, lyophilized |
4277 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
AEPHSLRYNL MVLSQDESVQ SGFLAEGHLD GQPFLRYDRQ KRRAKPQGQW AEDVLGAKTW DTETEDLTEN GQDLRRTLTH IKDQKGGLHS LQEIRVCEIH EDSSTRGSRH FYYDGELFLS QNLETQESTV PQSSRAQTLA MNVTNFWKED AMKTKTHYRA MQADCLQKLQ RYLKSGVAIR RTVPPMVNVT CSEVSEGNIT VTCRASSFYP RNITLTWRQD GVSLSHNTQQ WGDVLPDGNG TYQTWVATRI RQGEEQRFTC YMEHSGNHGT HPVPSGKVLV LQSQRTD | |
Greater than 95% as determined by SDS-PAGE and HPLC. |