missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MESDC1 Protein
A cDNA sequence encoding the MESDC1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12680-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
MESDC1 Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MESDC1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM DTT. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMASGSAG KPTGEAASPA PASAIGGASS QPRKRLVSVC DHCKGKMQLV ADLLLLSSEA RPVLFEGPAS SGAGAESFEQ CRDTIIARTK GLSILTHDVQ SQLNMGRFGE AGDSLVELGD LVVSLTECSA HAAYLAAVAT PGAQPAQPGL VDRYRVTRCR HEVEQGCAVL RATPLADMTP QLLLEVSQGL SRNLKFLTDA CALASDKSRD RFSREQFKLG VKCMSTSASA LLACVREVKV APSELARSRC ALFSGPLVQA VSALVGFATE PQFLGRAAAV SAEGKAVQTA ILGGAMSVVS ACVLLTQCLR DLAQHPDGGA KMSDHRERLR NSACAVSEGC TLLSQALRER SSPRTLPPVN SNSVN |