missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAPRE2 Protein
A cDNA sequence encoding the MAPRE2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12645-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
MAPRE2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPGPTQTLSP NGENNNDIIQ DNNGTIIPFR KHTVRGERSY SWGMAVNVYS TSITQETMSR HDIIAWVNDI VSLNYTKVEQ LCSGAAYCQF MDMLFPGCIS LKKVKFQAKL EHEYIHNFKL LQASFKRMNV DKVIPVEKLV KGRFQDNLDF IQWFKKFYDA NYDGKEYDPV EARQGQDAIP PPDPGEQIFN LPKKSHHANS PTAGAAKSSP AAKPGSTPSR PSSAKRASSS GSASKSDKDL ETQVIQLNEQ VHSLKLALEG VEKERDFYFG KLREIELLCQ EHGQENDDLV QRLMDILYAS EEHEGHTEEP EAEEQAHEQQ PPQQEEY | |
Greater than 85% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MAPRE2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |