missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MANF / ARMET Protein
A DNA sequence encoding the Human MANF / ARMET, was expressed in the hosts and tags indicated.
105.00€ - 1970.00€
Especificaciones
Número de acceso | NP_006001.4 |
---|---|
ID de gen (Entrez) | 7873 |
Nombre | MANF / ARMET Protein |
Estado normativo | Research Use Only |
Concentración de endotoxinas | Less than 1.0 EU/ug as determined by LAL method. |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15923798
|
enQuireBio™
QP10418-ec-5ug |
5 μg | |||||||
15913798
|
enQuireBio™
QP10418-ec-25ug |
25 μg | |||||||
15993788
|
enQuireBio™
QP10418-ec-100ug |
100 μg | |||||||
15903798
|
enQuireBio™
QP10418-ec-250ug |
250 μg | |||||||
Especificaciones
NP_006001.4 | |
MANF / ARMET Protein | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
LRPGDCEVCI SYLGRFYQDL KDRDVTFSPA TIENELIKFC REARGKENRL CYYIGATDDA ATKIINEVSK PLAHHIPVEK ICEKLKKKDS QICELKYDKQ IDLSTVDLKK LRVKELKKIL DDWGETCKGC AEKSDYIRKI NELMPKYAPK AASARTDL | |
Greater than 95% as determined by SDS-PAGE and HPLC. |
7873 | |
Research Use Only | |
MANF | |
The ED50 as determined by a cell proliferation assay using rat C6 cells is less than 20 ug/ml, corresponding to a specific activity of > 50 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |