missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Lipocalin-2/NGAL Protein

Product Code. 18653379 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
1000 μg
100 μg
10 μg
500 μg
Unit Size:
10 microgramos
100 microgramos
1000 microgramos
500 microgramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18653379 1000 μg 1000 microgramos
18687766 10 μg 10 microgramos
18603338 100 μg 100 microgramos
18673668 500 μg 500 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18653379 Supplier Novus Biologicals™ Supplier No. NBP2613291000ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Recombinant Protein

Lipocalin-2/NGAL protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 0.5 ng/mL, corresponding to a specific activity of > 2.0 x 10^6 IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

Para utilizar con (aplicación) Bioactivity
Formulación Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4, with 0.05 % Tween-20.
ID de gen (Entrez) 3934
Peso molecular MolecularWeight-theroretical: 41 kDa
Cantidad 1000 μg
Requisitos de almacenamiento Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Concentración de endotoxinas Less than 0.1 EU/μg of Lipocalin-2/NGAL as determined by LAL method.
Alias de gen 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin
Símbolo de gen LCN2
Reconstitución Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Categoría de investigación Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience
Grado de pureza o calidad >95%, by SDS-PAGE and HPLC
Expresión Lipocalin-2/NGAL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.