missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Keratin 36 GST (N-Term) Protein
Tienda Bio Techne Productos
Click to view available options
Cantidad:
10 μg
25 μg
missing translation for 'unitSize'
10 microgramos
25 microgramos
Beschreibung
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Spezifikation
Spezifikation
| Para utilizar con (aplicación) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulación | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| ID de gen (Entrez) | 8689 |
| Peso molecular | MolecularWeight-theroretical: 36.74 kDa |
| Cantidad | 25 μg |
| Requisitos de almacenamiento | Store at -80°C. Avoid freeze-thaw cycles. |
| Alias de gen | HA6, Hair keratin, type I Ha6, HHA6, HKA6, K36, keratin 36, keratin, hair, acidic, 6, keratin-36, KRTHA6keratin, type I cuticular Ha6 |
| Símbolo de gen | KRT36 |
| Categoría de investigación | Cell Biology, Cytoskeleton Markers, Signal Transduction |
| Grado de pureza o calidad | >80% by SDS-PAGE and Coomassie blue staining |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur