Learn More
enQuireBio™ Recombinant Human Intraflagellar transport protein 27 homolog Protein
A DNA sequence encoding the Homo sapiens (Human) Intraflagellar transport protein 27 homolog, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8131-ec-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001171172.1 | |
Intraflagellar transport protein 27 homolog Protein | |
Research Use Only | |
IFT27 | |
Recombinant Protein | |
E. coli | |
MVKLAAKCIL AGDPAVGKTA LAQIFRSDGA HFQKSYTLTT GMDLVVKTVP VPDTGDSVEL FIFDSAGKEL FSEMLDKLWE SPNVLCLVYD VTNEESFNNC SKWLEKARSQ APGISLPGVL VGNKTDLAGR RAVDSAEARA WALGQGLECF ETSVKEMENF EAPFHCLAKQ FHQLYREKVE VFRALA | |
Greater than 90% as determined by SDS-PAGE. |
11020 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |