Learn More
Novus Biologicals™ Recombinant Human IgA2 GST (N-Term) Protein
Código de producto. 18620551 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
10 μg
25 μg
Tamaño de la unidad:
10 microgramos
25 microgramos
18620551 25 μg 25 microgramos
18695151 10 μg 10 microgramos
2 options
Código de producto. 18620551
Marca: Novus Biologicals™ H00003494P0125ug
Recombinant Protein
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY
Especificaciones
Para utilizar con (aplicación) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulación 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
ID de gen (Entrez) 3494
Peso molecular MolecularWeight-theroretical: 78 kDa
Cantidad 25 μg
Requisitos de almacenamiento Store at -80°C. Avoid freeze-thaw cycles.
Alias de gen immunoglobulin heavy constant alpha 2 (A2m marker)
Símbolo de gen IGHA2
Categoría de investigación Immunology
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Expresión IgA2