missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beskrivelse
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Tekniske data
Tekniske data
| Para utilizar con (aplicación) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulación | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| ID de gen (Entrez) | 154214 |
| Peso molecular | MolecularWeight-theroretical: 36.63 kDa |
| Cantidad | 25 μg |
| Requisitos de almacenamiento | Store at -80°C. Avoid freeze/thaw cycles. |
| Alias de gen | C6orf172, chromosome 6 open reading frame 172, dJ84N20.1, EC 6.3.2.-, IBR domain containing 1, IBRDC1, MGC26996, probable E3 ubiquitin-protein ligase RNF217, ring finger protein 217 IBR domain-containing protein 1 |
| Símbolo de gen | RNF217 |
| Grado de pureza o calidad | >80% by SDS-PAGE and Coomassie blue staining |
| Expresión | IBRDC1 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?