missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HES2 Protein
A cDNA sequence encoding the HES2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12219-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
HES2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGLPRRA GDAAELRKSL KPLLEKRRRA RINQSLSQLK GLILPLLGRE NSNCSKLEKA DVLEMTVRFL QELPASSWPT AAPLPCDSYR EGYSACVARL ARVLPACRVL EPAVSARLLE HLWRRAASAT LDGGRAGDSS GPSAPAPAPA SAPEPASAPV PSPPSPPCGP GLWRPW | |
Greater than 80.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HES2 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.4M Urea and 10% glycerol. |