missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GZMH Protein
A cDNA sequence encoding the GZMH was constructed and used to recombinantly synthesize the protein.
Especificaciones
ID de gen (Entrez) | 2999 |
---|---|
Nombre | GZMH Protein |
Estado normativo | Research Use Only |
Concentración de endotoxinas | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Símbolo de gen | GZMH |
Especificaciones
2999 | |
Research Use Only | |
GZMH | |
Human | |
His | |
GZMH protein solution (0.25 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |
GZMH Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MGSSHHHHHH SSGLVPRGSH MEIIGGHEAK PHSRPYMAFV QFLQEKSRKR CGGILVRKDF VLTAAHCQGS SINVTLGAHN IKEQERTQQF IPVKRPIPHP AYNPKNFSND IMLLQLERKA KWTTAVRPLR LPSSKAQVKP GQLCSVAGWG YVSMSTLATT LQEVLLTVQK DCQCERLFHG NYSRATEICV GDPKKTQTGF KGDSGGPLVC KDVAQGILSY GNKKGTPPGV YIKVSHFLPW IKRTMKRL | |
Greater than 85.0% as determined by SDS-PAGE. |