missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FAM107B Protein
A cDNA sequence encoding the FAM107B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11842-20ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
FAM107B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAEPDYI EDDNPELIRP QKLINPVKTS RNHQDLHREL LMNQKRGLAP QNKPELQKVM EKRKRDQVIK QKEEEAQKKK SDLEIELLKR QQKLEQLELE KQKLQEEQEN APEFVKVKGN LRRTGQEVAQ AQES | |
Greater than 95% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The FAM107B solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |