missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Especificaciones
Specifikationer
| Para utilizar con (aplicación) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulación | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| ID de gen (Entrez) | 284611 |
| Peso molecular | MolecularWeight-theroretical: 62 kDa |
| Cantidad | 10 μg |
| Requisitos de almacenamiento | Store at -80°C. Avoid freeze/thaw cycles. |
| Alias de gen | DKFZp686N01110, DKFZp779B126, family with sequence similarity 102, member B, hypothetical protein LOC284611 |
| Símbolo de gen | FAM102B |
| Grado de pureza o calidad | >80% by SDS-PAGE and Coomassie blue staining |
| Expresión | FAM102B |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?