missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF3K Protein
A cDNA sequence encoding the EIF3K was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11769-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
EIF3K Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAMFEQMRAN VGKLLKGIDR YNPENLATLE RYVETQAKEN AYDLEANLAV LKLYQFNPAF FQTTVTAQIL LKALTNLPHT DFTLCKCMID QAHQEERPIR QILYLGDLLE TCHFQAFWQA LDENMDLLEG ITGFEDSVRK FICHVVGITY QHIDRWLLAE MLGDLSDSQL KVWMSKYGWS ADESGQIFIC SQEESIKPKN IVEKIDFDSV SSIMASSQ | |
Greater than 95% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The EIF3K protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 20% glycerol. |