missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF3J Protein
A cDNA sequence encoding the EIF3J was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11768-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
EIF3J Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MKISEKKKIA EKIKEKERQQ KKRQEEIKKR LEEPEEPKVL TPEEQLADKL RLKKLQEESD LELAKETFGV NNAVYGIDAM NPSSRDDFTE FGKLLKDKIT QYEKSLYYAS FLEVLVRDVC ISLEIDDLKK ITNSLTVLCS EKQKQEKQSK AKKKKKGVVP GGGLKATMKD DLADYGGYDG GYVQDYEDFM | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The EIF3J solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 10% glycerol and 200mM NaCl. |