missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Defensin alpha 3 GST (N-Term) Protein

Código de producto. 18682981 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
10 μg
25 μg
Unit Size:
10 microgramos
25 microgramos
Product Code. Cantidad unitSize
18682981 10 μg 10 microgramos
18643040 25 μg 25 microgramos
2 options
This item is not returnable. View return policy

Product Code. 18682981

Brand: Novus Biologicals™ H00001668P0110ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

Para utilizar con (aplicación) Western Blot, ELISA, SDS-PAGE, Protein Array, Immunoaffinity Purification
Formulación 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
ID de gen (Entrez) 1668
Peso molecular MolecularWeight-theroretical: 36.6 kDa
Cantidad 10 μg
Requisitos de almacenamiento Store at -80°C. Avoid freeze-thaw cycles.
Alias de gen DEF3, Defensin, alpha 3, defensin, alpha 3, neutrophil-specific, HNP3, HNP-3defensin 3, neutrophil-specific, HP3, HP-3, neutrophil defensin 3, neutrophil peptide 3
Símbolo de gen DEFA3
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Expresión Defensin alpha 3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.