missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CEBP Alpha Protein
A cDNA sequence encoding the CEBP Alpha was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11383-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CEBP Alpha Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMGAG KAKKSVDKNS NEYRVRRERN NIAVRKSRDK AKQRNVETQQ KVLELTSDND RLRKRVEQLS RELDTLRGIF RQLPESSLVKAMGNCA | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The protein contains 20mM Tris-HCl pH 7.5, 0.1M NaCl and 5mM b-Mercaptoethanol. |