missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD5L Protein
A cDNA sequence encoding the CD5L was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11341-HIS-HEK-10ug
Additional Details : Peso : 0.01000kg
Specifications
922 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human (HEK293) | |
MKHHHHHHAS PSGVRLVGGL HRCEGRVEVE QKGQWGTVCD DGWDIKDVAV LCRELGCGAA SGTPSGILYE PPAEKEQKVL IQSVSCTGTE DTLAQCEQEE VYDCSHDEDA GASCENPESS FSPVPEGVRL ADGPGHCKGR VEVKHQNQWY TVCQTGWSLR AAKVVCRQLG CGRAVLTQKR CNKHAYGRKP IWLSQMSCSG REATLQDCPS GPWGKNTCNH DEDTWVECED PFDLRLVGGD NLCSGRLEVL HKGVWGSVCD DNWGEKEDQV VCKQLGCGKS LSPSFRDRKC YGPGVGRIWL DNVRCSGEEQ SLEQCQHRFW GFHDCTHQED VAVICSG | |
Greater than 95.0% as determined by SDS-PAGE. |
CD5L Protein | |
Research Use Only | |
CD5L | |
Human | |
His | |
CD5L filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 50mM Tris buffer, pH 8.6. |