missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human CCL5/RANTES Protein

Código de producto. 18647515 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
1 mg
500 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
5 microgramos
500 microgramos
Código de producto. Cantidad unitSize
18647515 500 μg 500 microgramos
18655245 100 μg 100 microgramos
18607835 1 mg 1 miligramo
18655805 5 μg 5 microgramos
4 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18647515

Marca: Novus Biologicals™ NBP235235500ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

RANTES Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 1.0-10 ng/mL.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P13501, P13501, P13501
Para utilizar con (aplicación) SDS-PAGE, Bioactivity
Formulación Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM PB, pH 7.4, 100 mM NaCl.
ID de gen (Entrez) 6352
Peso molecular MolecularWeight-theroretical: 7.8 kDa
Cantidad 500 μg
Requisitos de almacenamiento Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Concentración de endotoxinas Less than 1 EU/μg of CCL5/RANTES as determined by LAL method.
Alias de gen CCL5, chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, RANTES/CCL5, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES
Símbolo de gen CCL5
Reconstitución Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Categoría de investigación Biologically Active Proteins, Chemokines and Cytokines
Grado de pureza o calidad >98 % pure by SDS-PAGE and HPLC
Expresión CCL5/RANTES
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.