Learn More
Novus Biologicals™ Recombinant Human ANKRD36 GST (N-Term) Protein
Código de producto. 18543394 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
10 μg
25 μg
Tamaño de la unidad:
10 microgramos
25 microgramos
18543394 10 μg 10 microgramos
18514684 25 μg 25 microgramos
2 options
Código de producto. 18543394
Marca: Novus Biologicals™ H00375248P0110ug
Recombinant Protein
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY
Especificaciones
Para utilizar con (aplicación) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulación 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
ID de gen (Entrez) 375248
Peso molecular MolecularWeight-theroretical: 45.3 kDa
Cantidad 10 μg
Requisitos de almacenamiento Store at -80°C. Avoid freeze/thaw cycles.
Alias de gen ankyrin repeat domain 36, ankyrin repeat domain-containing protein 36, UNQ2430
Símbolo de gen ANKRD36
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Expresión ANKRD36