Learn More
Novus Biologicals™ Recombinant Human Adrenomedullin R/ADMR/GPR182 GST (N-Term) Protein
Código de producto. p-200052776 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
10 μg
25 μg
Tamaño de la unidad:
10 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18343209 10 μg 10 microgramos
18490220 25 μg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18343209 Proveedor Novus Biologicals™ N.º de proveedor H00011318Q0110ug
Recombinant Protein
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY
Especificaciones
Para utilizar con (aplicación) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulación 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
ID de gen (Entrez) 11318
Peso molecular 31.68 kDa
Cantidad 10 μg
Requisitos de almacenamiento Store at -80°C. Avoid freeze-thaw cycles.
Alias de gen ADMR, adrenomedullin receptor, AMR, AM-R, G protein-coupled receptor 182,7TMR, G10D, gamrh, G-protein coupled receptor 182, hrhAMR, MGC34399
Símbolo de gen GPR182
Categoría de investigación GPCR
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Expresión Adrenomedullin R/ADMR/GPR182