missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADAP1 Protein
A DNA sequence encoding the Homo sapiens (Human) ADAP1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5612-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001271237.1 | |
ADAP1 Protein | |
Research Use Only | |
ADAP1 | |
Recombinant Protein | |
E. coli | |
MAKERRRAVL ELLQRPGNAR CADCGAPDPD WASYTLGVFI CLSCSGIHRN IPQVSKVKSV RLDAWEEAQV EFMASHGNDA ARARFESKVP SFYYRPTPSD CQLLREQWIR AKYERQEFIY PEKQEPYSAG YREGFLWKRG RDNGQFLSRK FVLTEREGAL KYFNRNDAKE PKAVMKIEHL NATFQPAKIG HPHGLQVTYL KDNSTRNIFI YHEDGKEIVD WFNALRAARF HYLQVAFPGA SDADLVPKLS RNYLKEGYME KTGPKQTEGF RKRWFTMDDR RLMYFKDPLD AFARGEVFIG SKESGYTVLH GFPPSTQGHH WPHGITIVTP DRKFLFACET ESDQREWVAA FQKAVDRPML PQEYAVEAHF KHKP | |
Greater than 80% as determined by SDS-PAGE. |
11033 | |
500 μg | |
Not determined. | |
Full Length of BC033747 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |