missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HSV HCV NS3 47.8kDa Protein
A cDNA sequence encoding the HCV NS3 47.8kDa was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12163-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
HSV HCV NS3 47.8kDa Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lyophilized from 1 mg/ml in 20mM sodium carbonate pH 9.6. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HSV | |
His | |
Greater than 95.0% as determined by SDS-PAGE. |