missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS3 Genotype-1b Protein
A cDNA sequence encoding the HCV NS3 Genotype-1b was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12165-HIS-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Hepatitis C HCV NS3 Genotype-1b Protein | |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
His | |
50mM NaPO4 pH 8.3 and 10mM DTT. |
HCV NS3 Genotype-1b protein is >95% pure as determined by 10% PAGE (coomassie staining). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Starts: MRDSDSQTFQ Ends: DSVIDCNTCVT |