missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli LptA Protein
A DNA sequence encoding the Escherichia coli (strain K12) LptA, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7331-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Specifica
NP_417667.1 | |
LptA Protein | |
Research Use Only | |
lptA | |
Recombinant Protein | |
E. coli | |
VTGDTDQPIH IESDQQSLDM QGNVVTFTGN VIVTQGTIKI NADKVVVTRP GGEQGKEVID GYGKPATFYQ MQDNGKPVEG HASQMHYELA KDFVVLTGNA YLQQVDSNIK GDKITYLVKE QKMQAFSDKG KRVTTVLVPS QLQDKNNKGQ TPAQKKGN | |
Greater than 90% as determined by SDS-PAGE. |
947920 | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |