Learn More
RBM4B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15412-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
May play a role in alternative splice site selection during pre-mRNA processing.Especificaciones
RBM4B | |
Polyclonal | |
Unconjugated | |
RBM4B | |
RBM30, RBM4B, RBM4L, RNA binding motif protein 30, RNA binding motif protein 4B, RNA binding protein 30, RNA binding protein 4B, ZCCHC15, ZCRB3B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
474154, 66704, 83759 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q64LC9, Q8VE92, Q9BQ04 | |
Rbm4b | |
RBM4B Fusion Protein Ag7679 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |